missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(2-pyrazinyl)-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

$136.63 - $136.63

Identifiants chimiques

CAS 216959-92-1
Molecular Formula C9H7N3O2S
Molecular Weight (g/mol) 221.23
MDL Number MFCD00111662
InChI Key XDZXQDHRHGIPRN-UHFFFAOYSA-N
Synonym 4-methyl-2-2-pyrazinyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid, 2-5-carboxy-4-methyl-1,3-thiazol-2-yl pyrazine, 4-methyl-2-2-pyrazinyl thiazole-5-carboxylic acid, 4-methyl-2-2-pyrazinyl-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl thiazole-5-carboxylic acid, 5-carboxy-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 5-thiazolecarboxylicacid, 4-methyl-2-2-pyrazinyl, 5-thiazolecarboxylic acid, 4-methyl-2-2-pyrazinyl
PubChem CID 2776505
IUPAC Name 4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C1=NC=CN=C1)C(O)=O
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
CC19801CB
FDS Afficher les documents
Thermo Scientific Chemicals
CC19801CB
250 mg
chaque for $136.63
Il en reste null
 
Description

Description

Spécifications

Identifiants chimiques

216959-92-1
221.23
XDZXQDHRHGIPRN-UHFFFAOYSA-N
2776505
CC1=C(SC(=N1)C1=NC=CN=C1)C(O)=O
C9H7N3O2S
MFCD00111662
4-methyl-2-2-pyrazinyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid, 2-5-carboxy-4-methyl-1,3-thiazol-2-yl pyrazine, 4-methyl-2-2-pyrazinyl thiazole-5-carboxylic acid, 4-methyl-2-2-pyrazinyl-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl thiazole-5-carboxylic acid, 5-carboxy-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 5-thiazolecarboxylicacid, 4-methyl-2-2-pyrazinyl, 5-thiazolecarboxylic acid, 4-methyl-2-2-pyrazinyl
4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid

Spécifications

216959-92-1
100.0
C9H7N3O2S
250 mg
XDZXQDHRHGIPRN-UHFFFAOYSA-N
4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
2776505
97%
96.5
97%
MFCD00111662
4-methyl-2-2-pyrazinyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid, 2-5-carboxy-4-methyl-1,3-thiazol-2-yl pyrazine, 4-methyl-2-2-pyrazinyl thiazole-5-carboxylic acid, 4-methyl-2-2-pyrazinyl-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl thiazole-5-carboxylic acid, 5-carboxy-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 5-thiazolecarboxylicacid, 4-methyl-2-2-pyrazinyl, 5-thiazolecarboxylic acid, 4-methyl-2-2-pyrazinyl
CC1=C(SC(=N1)C1=NC=CN=C1)C(O)=O
221.23
221.24
4-Methyl-2-(2-pyrazinyl)-1, 3-thiazole-5-carboxylic acid
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

FDS
missing translation for 'documents'

missing translation for 'documents'