missing translation for 'onlineSavingsMsg'
Learn More

4'-Iodoacetophenone, 98%

CAS: 13329-40-3 | C8H7IO | 246.047 g/mol

Supplier:  Thermo Scientific Chemicals A1679414

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA1679414


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions

Solubility
Easily soluble in water.

Notes
Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Spécifications

Spécifications

4'-Iodoacetophenone
82°C to 84°C
MFCD00045320
1857412
4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone
JZJWCDQGIPQBAO-UHFFFAOYSA-N
1-(4-iodophenyl)ethanone
72869
98%
13329-40-3
C8H7IO
25 g
Light sensitive
Easily soluble in water.
CC(=O)C1=CC=C(C=C1)I
246.047
246.05

Identifiants chimiques

13329-40-3
246.047
JZJWCDQGIPQBAO-UHFFFAOYSA-N
72869
CC(=O)C1=CC=C(C=C1)I
C8H7IO
MFCD00045320
4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone
1-(4-iodophenyl)ethanone
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 236-372-8

TSCA : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only