missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorothiophenol, 97%
Description
It is used in the synthesis of Bicalutamide, an anti-cancer drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the synthesis of Bicalutamide, an anti-cancer drug.
Solubility
Not miscible or difficult to mix in water.
Notes
Air Sensitive. Keep away from oxidizing agents and air. Store under inert gas.
It is used in the synthesis of Bicalutamide, an anti-cancer drug.
Solubility
Not miscible or difficult to mix in water.
Notes
Air Sensitive. Keep away from oxidizing agents and air. Store under inert gas.
Spécifications
Spécifications
| Density | 1.203 |
| Boiling Point | 170°C to 172°C |
| Flash Point | 57°C (135°F) |
| Odor | Unpleasant |
| Refractive Index | 1.55 |
| Quantity | 5 g |
| UN Number | UN3071 |
| Beilstein | 1446350 |
| Sensitivity | Air Sensitive |
| Solubility Information | Not miscible or difficult to mix in water. |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H226-H301+H311+H331-H315-H319-H335 |
| Hazard Statement | GHS H Statement H300-H310-H331-H226-H315-H319-H317-H335 Fatal if swallowed. Fatal in contact with skin. Toxic if inhaled. Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: III |
| EINECSNumber | 206-737-6 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only