Learn More
4-Bromophenylboronic acid, 98%, Thermo Scientific™
Identifiants chimiques
| CAS | 5467-74-3 |
|---|---|
| Molecular Formula | C6H6BBrO2 |
| Molecular Weight (g/mol) | 200.83 |
| MDL Number | MFCD00002104 |
| InChI Key | QBLFZIBJXUQVRF-UHFFFAOYSA-N |
| Synonym | 4-bromophenyl boronic acid, 4-bromobenzeneboronic acid, 4-bromophenylboric acid, p-bromophenylboronic acid, p-bromophenylboric acid, p-bromobenzeneboronic acid, boronic acid, 4-bromophenyl, benzeneboronic acid, p-bromo, p-bromophenyl boronic acid, 4-bromo phenyl boronic acid |
| PubChem CID | 79599 |
| IUPAC Name | (4-bromophenyl)boronic acid |
| SMILES | B(C1=CC=C(C=C1)Br)(O)O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC107280050
|
Thermo Scientific Chemicals
107280050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
|
AC107280250
|
Thermo Scientific Chemicals
107280250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
| 5467-74-3 | |
| 200.83 | |
| QBLFZIBJXUQVRF-UHFFFAOYSA-N | |
| 79599 | |
| B(C1=CC=C(C=C1)Br)(O)O |
| C6H6BBrO2 | |
| MFCD00002104 | |
| 4-bromophenyl boronic acid, 4-bromobenzeneboronic acid, 4-bromophenylboric acid, p-bromophenylboronic acid, p-bromophenylboric acid, p-bromobenzeneboronic acid, boronic acid, 4-bromophenyl, benzeneboronic acid, p-bromo, p-bromophenyl boronic acid, 4-bromo phenyl boronic acid | |
| (4-bromophenyl)boronic acid |
Spécifications
| 5467-74-3,5467-74-3 | |
| 100.0 | |
| 97.5% min. (HPLC) | |
| C6H6BBrO2 | |
| MFCD00002104 | |
| 16, II, 638 | |
| Solubility in water: .. Other solubilities: soluble in ether,thf,dmso,dmf,methanol | |
| B(C1=CC=C(C=C1)Br)(O)O | |
| 200.83 | |
| 200.83 | |
| 4-Bromophenylboronic acid |
| 97.5 | |
| Authentic | |
| Glass bottle | |
| BrC6H4B(OH)2 | |
| 5g | |
| 4-bromophenyl boronic acid, 4-bromobenzeneboronic acid, 4-bromophenylboric acid, p-bromophenylboronic acid, p-bromophenylboric acid, p-bromobenzeneboronic acid, boronic acid, 4-bromophenyl, benzeneboronic acid, p-bromo, p-bromophenyl boronic acid, 4-bromo phenyl boronic acid | |
| QBLFZIBJXUQVRF-UHFFFAOYSA-N | |
| (4-bromophenyl)boronic acid | |
| 79599 | |
| 98% |
Sécurité et manipulation
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
If skin irri
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 226-779-9
RUO â Research Use Only