missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 97%

CAS: 874838-96-7 | C12H7ClF3NO3S | 337.70 g/mol

Supplier:  Thermo Scientific Chemicals H3328303

 Afficher plus de versions de ce produit

Numéro de catalogue. AAH3328303

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride
51°C to 53°C
MFCD07774162
UN3261
XEMAYRPWJCLKBQ-UHFFFAOYSA-N
4-[5-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
24213828
97%
874838-96-7
C12H7ClF3NO3S
1 g
4-5-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-5-trifluoromethyl-2-pyridinyl oxy benzenesulfonoyl chloride, 4-5-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, 4-5-trifluoromethyl-2-pyridinyl-oxy benzenesulfonoyl chloride, 4-5-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride, trifluoromethylpyridinyloxybenzenesulfonoylchloride, 4-5-trifluoromethyl-2-pyridyloxy benzenesulfonyl chloride, chloro 4-5-trifluoromethyl 2-pyridyloxy phenyl sulfone
FC(F)(F)C1=CN=C(OC2=CC=C(C=C2)S(Cl)(=O)=O)C=C1
337.70
337.71

Identifiants chimiques

874838-96-7
337.70
XEMAYRPWJCLKBQ-UHFFFAOYSA-N
24213828
FC(F)(F)C1=CN=C(OC2=CC=C(C=C2)S(Cl)(=O)=O)C=C1
C12H7ClF3NO3S
MFCD07774162
4-5-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-5-trifluoromethyl-2-pyridinyl oxy benzenesulfonoyl chloride, 4-5-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, 4-5-trifluoromethyl-2-pyridinyl-oxy benzenesulfonoyl chloride, 4-5-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride, trifluoromethylpyridinyloxybenzenesulfonoylchloride, 4-5-trifluoromethyl-2-pyridyloxy benzenesulfonyl chloride, chloro 4-5-trifluoromethyl 2-pyridyloxy phenyl sulfone
4-[5-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only