missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylpiperazino)benzylamine, ≥90%, Thermo Scientific™

Identifiants chimiques

CAS 216144-45-5
Molecular Formula C12H19N3
Molecular Weight (g/mol) 205.305
MDL Number MFCD03086117
InChI Key MZFQGKRIWIKPBT-UHFFFAOYSA-N
Synonym 4-4-methylpiperazino benzylamine, 4-4-methylpiperazin-1-yl benzylamine, 4-4-methylpiperazin-1-yl phenyl methanamine, 1-4-4-methylpiperazin-1-yl phenyl methanamine, 4-4-methyl-piperazin-1-yl-benzylamine, 4-4-methylpiperazin-1-yl-benzylamine, benzenemethanamine, 4-4-methyl-1-piperazinyl, 4-4-methylpiperazinyl phenyl methylamine, pubchem9053
PubChem CID 2776493
IUPAC Name [4-(4-methylpiperazin-1-yl)phenyl]methanamine
SMILES CN1CCN(CC1)C2=CC=C(C=C2)CN
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
CC18713CB
FDS Afficher les documents
Thermo Scientific Chemicals
CC18713CB
250mg Amber Glass Bottle N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
CC18713DA
FDS Afficher les documents
Thermo Scientific Chemicals
CC18713DA
1g Amber Glass Bottle N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Description

Description

Spécifications

Identifiants chimiques

216144-45-5
205.305
MZFQGKRIWIKPBT-UHFFFAOYSA-N
2776493
CN1CCN(CC1)C2=CC=C(C=C2)CN
C12H19N3
MFCD03086117
4-4-methylpiperazino benzylamine, 4-4-methylpiperazin-1-yl benzylamine, 4-4-methylpiperazin-1-yl phenyl methanamine, 1-4-4-methylpiperazin-1-yl phenyl methanamine, 4-4-methyl-piperazin-1-yl-benzylamine, 4-4-methylpiperazin-1-yl-benzylamine, benzenemethanamine, 4-4-methyl-1-piperazinyl, 4-4-methylpiperazinyl phenyl methylamine, pubchem9053
[4-(4-methylpiperazin-1-yl)phenyl]methanamine

Spécifications

216144-45-5
100.0
Amber Glass Bottle
MFCD03086117
4-4-methylpiperazino benzylamine, 4-4-methylpiperazin-1-yl benzylamine, 4-4-methylpiperazin-1-yl phenyl methanamine, 1-4-4-methylpiperazin-1-yl phenyl methanamine, 4-4-methyl-piperazin-1-yl-benzylamine, 4-4-methylpiperazin-1-yl-benzylamine, benzenemethanamine, 4-4-methyl-1-piperazinyl, 4-4-methylpiperazinyl phenyl methylamine, pubchem9053
CN1CCN(CC1)C2=CC=C(C=C2)CN
205.305
205.3
4-(4-Methylpiperazino)benzylamine
89.5
90%
C12H19N3
250mg
MZFQGKRIWIKPBT-UHFFFAOYSA-N
[4-(4-methylpiperazin-1-yl)phenyl]methanamine
2776493
90%
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

FDS
missing translation for 'documents'

missing translation for 'documents'