missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Diaminostilbene-2,2'-disulfonic Acid 94.0+%, TCI America™
Supplier: TCI America D0121100G
Spécifications
| 4,4′-Diaminostilbene-2,2′-disulfonic Acid | |
| C14H14N2O6S2 | |
| 100 g | |
| REJHVSOVQBJEBF-OWOJBTEDSA-N | |
| 5-amino-2-[(E)-2-(4-amino-2-sulfophenyl)ethenyl]benzenesulfonic acid | |
| 5284378 | |
| ≥94.0% (T) |
| 81-11-8 | |
| MFCD00024946 | |
| amsonic acid, flavonic acid, tinopal bhs, amsonsaeure, dasd, 4,4'-diaminostilbene-2,2'-disulfonic acid, unii-h68088wvjl, 2,2'-disulfo-4,4'-stilbenediamine, diaminostilbenedisulfonic acid, 4,4'-diaminostilbene-2,2'-disulphonic acid | |
| C1=CC(=C(C=C1N)S(=O)(=O)O)C=CC2=C(C=C(C=C2)N)S(=O)(=O)O | |
| 370.394 | |
| 370.39 | |
| Crystalline Powder |
Identifiants chimiques
| 81-11-8 | |
| 370.394 | |
| REJHVSOVQBJEBF-OWOJBTEDSA-N | |
| 5284378 | |
| C1=CC(=C(C=C1N)S(=O)(=O)O)C=CC2=C(C=C(C=C2)N)S(=O)(=O)O |
| C14H14N2O6S2 | |
| MFCD00024946 | |
| amsonic acid, flavonic acid, tinopal bhs, amsonsaeure, dasd, 4,4'-diaminostilbene-2,2'-disulfonic acid, unii-h68088wvjl, 2,2'-disulfo-4,4'-stilbenediamine, diaminostilbenedisulfonic acid, 4,4'-diaminostilbene-2,2'-disulphonic acid | |
| 5-amino-2-[(E)-2-(4-amino-2-sulfophenyl)ethenyl]benzenesulfonic acid |
Sécurité et manipulation
missing translation for 'einecsNumber' : (4)-0288
missing translation for 'rtecsNumber' : WJ6603000
missing translation for 'tsca' : Yes