missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(2-methyl-3-furyl)-1,3,2-dioxaborolane, 90%, Thermo Scientific™

$182.69 - $182.69

Identifiants chimiques

CAS 864776-02-3
Molecular Formula C11H17BO3
Molecular Weight (g/mol) 208.06
MDL Number MFCD09879905
InChI Key JMLBPHNESOKSEV-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-2-methylfuran-3-yl-1,3,2-dioxaborolane, 2-methylfuran-3-boronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-2-methyl-3-furanyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane,4,4,5,5-tetramethyl-2-2-methyl-3-furanyl, 4,4,5,5-tetramethyl-2-2-methyl-3-furyl-1,3,2-dioxaborolane, pubchem18438, 2-2-methylfur-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan, 4,4,5,5-tetramethyl-2-2-methyl 3-furyl-1,3,2-dioxaborolane, 3-hydroxy-2,3-dimethylbutan-2-yl hydrogen 2-methylfuran-3-yl boronate
PubChem CID 24229547
IUPAC Name 4,4,5,5-tetramethyl-2-(2-methylfuran-3-yl)-1,3,2-dioxaborolane
SMILES CC1=C(C=CO1)B1OC(C)(C)C(C)(C)O1
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
CC55439CB
FDS Afficher les documents
Thermo Scientific Chemicals
CC55439CB
250 mg Amber Glass Bottle
chaque for $182.69
Il en reste null
 
Description

Description

Spécifications

Identifiants chimiques

864776-02-3
208.06
JMLBPHNESOKSEV-UHFFFAOYSA-N
24229547
CC1=C(C=CO1)B1OC(C)(C)C(C)(C)O1
C11H17BO3
MFCD09879905
4,4,5,5-tetramethyl-2-2-methylfuran-3-yl-1,3,2-dioxaborolane, 2-methylfuran-3-boronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-2-methyl-3-furanyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane,4,4,5,5-tetramethyl-2-2-methyl-3-furanyl, 4,4,5,5-tetramethyl-2-2-methyl-3-furyl-1,3,2-dioxaborolane, pubchem18438, 2-2-methylfur-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan, 4,4,5,5-tetramethyl-2-2-methyl 3-furyl-1,3,2-dioxaborolane, 3-hydroxy-2,3-dimethylbutan-2-yl hydrogen 2-methylfuran-3-yl boronate
4,4,5,5-tetramethyl-2-(2-methylfuran-3-yl)-1,3,2-dioxaborolane

Spécifications

864776-02-3
100.0
Amber Glass Bottle
MFCD09879905
4,4,5,5-tetramethyl-2-2-methylfuran-3-yl-1,3,2-dioxaborolane, 2-methylfuran-3-boronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-2-methyl-3-furanyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane,4,4,5,5-tetramethyl-2-2-methyl-3-furanyl, 4,4,5,5-tetramethyl-2-2-methyl-3-furyl-1,3,2-dioxaborolane, pubchem18438, 2-2-methylfur-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan, 4,4,5,5-tetramethyl-2-2-methyl 3-furyl-1,3,2-dioxaborolane, 3-hydroxy-2,3-dimethylbutan-2-yl hydrogen 2-methylfuran-3-yl boronate
CC1=C(C=CO1)B1OC(C)(C)C(C)(C)O1
208.06
208.06
4,4,5,5-Tetramethyl-2-(2-methyl-3-furyl)-1,3,2-dioxaborolane
89.5
90%
C11H17BO3
250 mg
JMLBPHNESOKSEV-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-(2-methylfuran-3-yl)-1,3,2-dioxaborolane
24229547
90%
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

FDS
missing translation for 'documents'

missing translation for 'documents'