missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ 7500 Series Reusable Half Face Mask Respirator

State-of-the-art features provide unsurpassed comfort. The 3M Reusable Half Face Mask Respirator uses a Bayonet Attachment System for twin lightweight filters to protect against gases, vapors, and particulates.

$37.19 - $38.30

Specifications

Certifications/Compliance NIOSH 42 CFR Part 84
Connection Type Bayonet
Product Type Half Mask Respirator
For Use With (Equipment) Cartridge 6000 series, filter 2000, 2200, 7000 or 5000 series, or dual airline system
Material Silicone
View More Specs

Requires: Compatible cartridges and/or filters. Accommodates 3M 7093 and Series 2000 filters and Series 6000 cartridges.


Products 3
Catalog Number Mfr. No. Size For Use With (Application) Price Quantity  
Catalog Number Mfr. No. Size For Use With (Application) Price Quantity  
19-096-898
Accessories View Documents
3M™
7100134948
Small Assembly, cleaning, grinding, machine operation, painting, remediation, welding
Each for $37.19
Case of 10 Each for $345.30
 
19-096-899
Accessories View Documents
3M™
7000002162
Medium Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
Each for $38.30
Case of 10 Each for $355.60
 
19-096-900
Accessories View Documents
3M™
7100134949
Large Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
Each for $38.30
Case of 10 Each for $355.60
 
Description

Description

Durable, reusable and heat-resistant mask is made of advanced silicone material that feels softer on the face, while the unique adjustment design helps reduce tension and pressure points on the face. 

  • Cool Flow™ exhalation valve opens easier, lowering exhalation breathing resistance, reducing heat and moisture buildup in the facepiece
  • Easily adjusted, dual-mode head harness gives user the option of wearing the respirator in either the traditional or drop-down mode
  • Yoke won't get hot like metal yokes, so the 7500 is an excellent choice for welding applications
  • Color-coded sizes for easy identification
  • 3M Catalog# 7501 , 3M Catalog# 7502 , 3M Catalog# 7503

 These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Compliance

NIOSH approved under 42 CFR 84 standard for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters; in Canada, CSA standard Z94.4 and applicable local government requirements must be met for approval.

Specifications

Specifications

NIOSH 42 CFR Part 84
Half Mask Respirator
Silicone
Bayonet
Cartridge 6000 series, filter 2000, 2200, 7000 or 5000 series, or dual airline system
SDS