missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-p-anisaldehyde 98.0+%, TCI America™

Numéro de catalogue. p-7133989
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
N08305G 5 g
N083025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. N08305G Fournisseur TCI America Code fournisseur N08305G
Il en reste null

Identifiants chimiques

CAS 31680-08-7
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00126498
InChI Key YTCRQCGRYCKYNO-UHFFFAOYSA-N
Synonym 3-nitro-4-anisaldehyde, 4-methoxy-3-nitro-benzaldehyde, benzaldehyde, 4-methoxy-3-nitro, 3-nitro-4-methoxybenzaldehyde, pubchem8145, 3-nitro-p-anisaldehyde, acmc-1adwt, 4-methoxy-5-nitrobenzaldehyde, 4-methoxy-3-nitrobenzaldehyde
PubChem CID 700608
IUPAC Name 4-methoxy-3-nitrobenzaldehyde
SMILES COC1=CC=C(C=O)C=C1[N+]([O-])=O

Spécifications

Melting Point 85°C
Color Yellow
Quantity 5 g
Formula Weight 181.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitro-p-anisaldehyde