missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butanol 98.0+%, TCI America™

Numéro de catalogue. p-7132033
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M017125ML 25 mL
M0171500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M017125ML Fournisseur TCI America Code fournisseur M017125ML
Il en reste null

Identifiants chimiques

CAS 598-75-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004527
InChI Key MXLMTQWGSQIYOW-UHFFFAOYNA-N
Synonym 3-methyl-2-butanol, sec-isoamyl alcohol, 2-butanol, 3-methyl, 2-methyl-3-butanol, isopropylmethylcarbinol, methylisopropylcarbinol, 1,2-dimethylpropanol, 1,2-dimethyl-1-propanol, dl-3-methyl-2-butanol, 3-methyl-butan-2-ol
PubChem CID 11732
ChEBI CHEBI:77517
IUPAC Name 3-methylbutan-2-ol
SMILES CC(C)C(C)O

Spécifications

Color Colorless
Boiling Point 112°C
Quantity 25 mL
UN Number 1105
Formula Weight 88.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-butanol
EINECSNumber (2)-0217
TSCA Yes