missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzaldehyde, 97%

CAS: 100-83-4 | C7H6O2 | 122.123 g/mol

Supplier:  Thermo Scientific Chemicals A1354122

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA1354122


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

3-Hydroxybenzaldehyde is used as an ionophore, during the development of an highly selective and sensitive PVC membrane sensor, which can be used as a Tb3+ ion selective electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzaldehyde is used as an ionophore, during the development of an highly selective and sensitive PVC membrane sensor, which can be used as a Tb3+ ion selective electrode.

Solubility
Soluble in DMSO and Methanol

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Spécifications

Spécifications

3-Hydroxybenzaldehyde
102°C to 105°C
C7H6O2
100 g
Air Sensitive
Soluble in DMSO and Methanol
C1=CC(=CC(=C1)O)C=O
122.123
CHEBI:16207
97%
100-83-4
191°C (50 mmHg)
MFCD00003368
507099
m-formylphenol, m-hydroxybenzaldehyde, 3-formylphenol, benzaldehyde, 3-hydroxy, meta-hydroxybenzaldehyde, benzaldehyde, m-hydroxy, 3-hydroxy-benzaldehyde, 3-hydroxy benzaldehyde, 3-hydroxybenzaldehyd, 3-hydroxylbenzaldehyde
IAVREABSGIHHMO-UHFFFAOYSA-N
3-hydroxybenzaldehyde
101
122.12

Identifiants chimiques

100-83-4
122.123
IAVREABSGIHHMO-UHFFFAOYSA-N
101
3-hydroxybenzaldehyde
C7H6O2
MFCD00003368
m-formylphenol, m-hydroxybenzaldehyde, 3-formylphenol, benzaldehyde, 3-hydroxy, meta-hydroxybenzaldehyde, benzaldehyde, m-hydroxy, 3-hydroxy-benzaldehyde, 3-hydroxy benzaldehyde, 3-hydroxybenzaldehyd, 3-hydroxylbenzaldehyde
CHEBI:16207
C1=CC(=CC(=C1)O)C=O
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

missing translation for 'einecsNumber' : 202-892-9

missing translation for 'rtecsNumber' : CU6450000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only