Learn More
3-Hydroxy-2-nitropyridine, 97%, Thermo Scientific™
$140.16 - $140.16
Identifiants chimiques
| CAS | 15128-82-2 |
|---|---|
| Molecular Formula | C5H4N2O3 |
| Molecular Weight (g/mol) | 140.10 |
| MDL Number | MFCD00006258 |
| InChI Key | QBPDSKPWYWIHGA-UHFFFAOYSA-N |
| Synonym | 3-hydroxy-2-nitropyridine, 2-nitropyridin-3-ol, 2-nitro-3-hydroxypyridine, 2-nitro-3-pyridinol, 3-pyridinol, 2-nitro, 2-nitro-3-hydroxy pyridine, unii-uu357x85yb, 3-hydroxy-2-nitro pyridine, 2-nitro-3-pyridol |
| PubChem CID | 27057 |
| IUPAC Name | 2-nitropyridin-3-ol |
| SMILES | OC1=CC=CN=C1[N+]([O-])=O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC148100250
|
Thermo Scientific Chemicals
148100250 |
25 g | Glass bottle |
chaque for $140.16
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Identifiants chimiques
| 15128-82-2 | |
| 140.10 | |
| QBPDSKPWYWIHGA-UHFFFAOYSA-N | |
| 27057 | |
| OC1=CC=CN=C1[N+]([O-])=O |
| C5H4N2O3 | |
| MFCD00006258 | |
| 3-hydroxy-2-nitropyridine, 2-nitropyridin-3-ol, 2-nitro-3-hydroxypyridine, 2-nitro-3-pyridinol, 3-pyridinol, 2-nitro, 2-nitro-3-hydroxy pyridine, unii-uu357x85yb, 3-hydroxy-2-nitro pyridine, 2-nitro-3-pyridol | |
| 2-nitropyridin-3-ol |
Spécifications
| 15128-82-2 | |
| 100.0 | |
| Yellow | |
| 97% | |
| C5H4N2O3 | |
| MFCD00006258 | |
| 3-hydroxy-2-nitropyridine, 2-nitropyridin-3-ol, 2-nitro-3-hydroxypyridine, 2-nitro-3-pyridinol, 3-pyridinol, 2-nitro, 2-nitro-3-hydroxy pyridine, unii-uu357x85yb, 3-hydroxy-2-nitro pyridine, 2-nitro-3-pyridol | |
| QBPDSKPWYWIHGA-UHFFFAOYSA-N | |
| 2-nitropyridin-3-ol | |
| 27057 | |
| 97% | |
| 3-Hydroxy-2-nitropyridine, 97% |
| 96.0 | |
| 67°C to 72°C | |
| Authentic | |
| Glass bottle | |
| HO(C5H3N)NO2 | |
| 25 g | |
| Solubility in water: soluble in water | |
| OC1=CC=CN=C1[N+]([O-])=O | |
| 140.10 | |
| 140.1 | |
| Crystalline Powder |
Sécurité et manipulation
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 239-191-2
missing translation for 'rtecsNumber' : UU7715000
missing translation for 'tsca' : TSCA
RUO – Research Use Only