missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromoimidazo[1,2-b]pyridazine 98.0+%, TCI America™
Supplier: TCI America B46521G
Spécifications
| 3-Bromoimidazo[1,2-b]pyridazine | |
| 150°C | |
| C6H4BrN3 | |
| 1 g | |
| KJQVHOFAWISYDO-UHFFFAOYSA-N | |
| 3-bromoimidazo[1,2-b]pyridazine | |
| 12872319 | |
| ≥98.0% (GC,T) |
| 18087-73-5 | |
| Yellow | |
| MFCD09757672 | |
| 3-bromoimidazo 1,2-b pyridazine, imidazo 1,2-b pyridazine, 3-bromo, pubchem18744, acmc-209eh2, ksc496a9t, 3-bromo-imidazo 1,2-b pyridazine, 3-bromoimidazo 1,2,-b pyridazine, 3-bromanylimidazo 1,2-b pyridazine, 3-bromo-4-hydroimidazo 1,2-e pyridazine | |
| BrC1=CN=C2C=CC=NN12 | |
| 198.02 | |
| 198.02 | |
| Crystalline Powder |
Identifiants chimiques
| 18087-73-5 | |
| 198.02 | |
| KJQVHOFAWISYDO-UHFFFAOYSA-N | |
| 12872319 | |
| BrC1=CN=C2C=CC=NN12 |
| C6H4BrN3 | |
| MFCD09757672 | |
| 3-bromoimidazo 1,2-b pyridazine, imidazo 1,2-b pyridazine, 3-bromo, pubchem18744, acmc-209eh2, ksc496a9t, 3-bromo-imidazo 1,2-b pyridazine, 3-bromoimidazo 1,2,-b pyridazine, 3-bromanylimidazo 1,2-b pyridazine, 3-bromo-4-hydroimidazo 1,2-e pyridazine | |
| 3-bromoimidazo[1,2-b]pyridazine |
Sécurité et manipulation
missing translation for 'tsca' : No