missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-5-mercapto-1,2,4-triazole 98.0+%, TCI America™
Supplier: TCI America A14555G
Spécifications
| 3-Amino-5-mercapto-1,2,4-triazole | |
| White-Yellow | |
| MFCD00005231 | |
| 3-amino-5-mercapto-1,2,4-triazole, iminothiourazole, 3-imino-5-thiourazole, 3-amino-1h-1,2,4-triazole-5-thiol, 5-amino-4h-1,2,4-triazole-3-thiol, 3-amino-1,2,4-triazole-5-thiol, s-triazole-3-thiol, 5-amino, 3h-1,2,4-triazole-3-thione, 5-amino-1,2-dihydro, 5-amino-1,2-dihydro-3h-1,2,4-triazole-3-thione, unii-353f2d8l1z | |
| NC1=NC(=S)NN1 | |
| 116.14 | |
| 116.14 | |
| Crystalline Powder |
| 16691-43-3 | |
| C2H4N4S | |
| 5 g | |
| WZUUZPAYWFIBDF-UHFFFAOYSA-N | |
| 5-amino-2,3-dihydro-1H-1,2,4-triazole-3-thione | |
| 2723869 | |
| ≥98.0% (HPLC,T) |
Identifiants chimiques
| 16691-43-3 | |
| 116.14 | |
| WZUUZPAYWFIBDF-UHFFFAOYSA-N | |
| 2723869 | |
| NC1=NC(=S)NN1 |
| C2H4N4S | |
| MFCD00005231 | |
| 3-amino-5-mercapto-1,2,4-triazole, iminothiourazole, 3-imino-5-thiourazole, 3-amino-1h-1,2,4-triazole-5-thiol, 5-amino-4h-1,2,4-triazole-3-thiol, 3-amino-1,2,4-triazole-5-thiol, s-triazole-3-thiol, 5-amino, 3h-1,2,4-triazole-3-thione, 5-amino-1,2-dihydro, 5-amino-1,2-dihydro-3h-1,2,4-triazole-3-thione, unii-353f2d8l1z | |
| 5-amino-2,3-dihydro-1H-1,2,4-triazole-3-thione |
Sécurité et manipulation
missing translation for 'einecsNumber' : (5)-0607
missing translation for 'rtecsNumber' : XZ5357500
missing translation for 'tsca' : Yes