missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylindole 98.0+%, TCI America™

Numéro de catalogue. p-7117086
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A08495G 5 g
A084925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A08495G Fournisseur TCI America Code fournisseur A08495G
Il en reste null

Identifiants chimiques

CAS 703-80-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00005626
InChI Key VUIMBZIZZFSQEE-UHFFFAOYSA-N
Synonym 3-acetylindole, 1-1h-indol-3-yl ethanone, 3-acetyl-1h-indole, ethanone, 1-1h-indol-3-yl, acetyl-3-indole, 1-1h-indol-3-yl-ethanone, ketone, indol-3-yl methyl, 3-acetyl indole, indol-3-yl methyl ketone, 1-1h-indol-3-yl ethan-1-one
PubChem CID 12802
IUPAC Name 1-(1H-indol-3-yl)ethanone
SMILES CC(=O)C1=CNC2=CC=CC=C21

Spécifications

Melting Point 190°C
Quantity 5 g
Formula Weight 159.19
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetylindole
RTECSNumber OB4520000
TSCA No