missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dihydroxybenzidine 99.0+%, TCI America™

Numéro de catalogue. d23125g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Numéro de catalogue. Quantity
D23125G 5 g
D231225G 25 g
2 options

Numéro de catalogue. D23125G

Supplier: TCI America D23125G

Il en reste null

Identifiants chimiques

CAS 2373-98-0
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD00039149
InChI Key ZGDMDBHLKNQPSD-UHFFFAOYSA-N
Synonym Benzidine-3,3′C-diol, 4,4′C-Diamino-3,3′C-dihydroxybiphenyl
PubChem CID 16918
IUPAC Name 4,4'-diamino-[1,1'-biphenyl]-3,3'-diol
SMILES NC1=CC=C(C=C1O)C1=CC=C(N)C(O)=C1

Spécifications

Melting Point 292°C
Quantity 5 g
Formula Weight 216.24
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dihydroxybenzidine
RTECSNumber DV4900000
TSCA No