missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ 2XYT-Broth, Capsules: Regular
Optimized for the growth and maintenance of M13 phage or other filamentous ssDNA bacteriophages
Supplier: MP Biomedicals™ 113012021
Description
Regular size capsules.
- Contents/L: 16g tryptone, 10g yeast extract, 5g NaCl
- Use 31 capsules/L
Spécifications
| Molecular Biology Certified™ | |
| Capsule | |
| Tryptone 16g/L, Yeast extract 10g/L, NaCl 5g/L | |
| 454 g |
| 15°C to 30°C | |
| Microbiology Media | |
| For growth and maintenance of M13 phage or other filamentous phage |