missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Utility Cart with Wired Shelves, Chrome
Carts consist of wire shelves with plastic split sleeves, two one-piece handles, donut bumpers, and casters.
$519.25 - $745.84
Specifications
| Product Type | Wire Utility Cart |
|---|---|
| Autoclavable | Not Autoclavable |
| Color | Silver |
| For Use With (Application) | Storage and Transport |
| Height (English) | 39 in. |
| Catalog Number | Mfr. No. | No. of Shelves | Length (English) | Width (English) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Shelves | Length (English) | Width (English) | Price | Quantity & Availability | |||
|
13460183
|
Fisherbrand™
FS1830NC2UC |
2 | 30 in. | 18 in. |
Each for $542.73
|
|
|||
|
13460187
|
Fisherbrand™
FS2430NC2UC |
2 | 30 in. | 24 in. |
Each for $588.96
|
|
|||
|
13460184
|
Fisherbrand™
FS1836NC2UC |
2 | 36 in. | 18 in. |
Each for $542.73
|
|
|||
|
13460188
|
Fisherbrand™
FS2436NC2UC |
2 | 36 in. | 24 in. |
Each for $588.96
|
|
|||
|
13460185
|
Fisherbrand™
FS1842NC2UC |
2 | 42 in. | 18 in. |
Each for $576.83
|
|
|||
|
13460189
|
Fisherbrand™
FS2442NC2UC |
2 | 42 in. | 24 in. |
Each for $519.25
|
|
|||
|
13460186
|
Fisherbrand™
FS1848NC2UC |
2 | 48 in. | 18 in. |
Each for $576.83
|
|
|||
|
13460190
|
Fisherbrand™
FS2448NC2UC |
2 | 48 in. | 24 in. |
Each for $623.11
|
|
|||
|
13460191
|
Fisherbrand™
FS1830NC3UC |
3 | 30 in. | 18 in. |
Each for $625.30
|
|
|||
|
13460195
|
Fisherbrand™
FS2430NC3UC |
3 | 30 in. | 24 in. |
Each for $694.65
|
|
|||
|
13460192
|
Fisherbrand™
FS1836NC3UC |
3 | 36 in. | 18 in. |
Each for $625.30
|
|
|||
|
13460196
|
Fisherbrand™
FS2436NC3UC |
3 | 36 in. | 24 in. |
Each for $694.65
|
|
|||
|
13460193
|
Fisherbrand™
FS1842NC3UC |
3 | 42 in. | 18 in. |
Each for $676.47
|
|
|||
|
13460197
|
Fisherbrand™
FS2442NC3UC |
3 | 42 in. | 24 in. |
Each for $745.84
|
|
|||
|
13460194
|
Fisherbrand™
FS1848NC3UC |
3 | 48 in. | 18 in. |
Each for $676.47
|
|
|||
|
13460198
|
Fisherbrand™
FS2448NC3UC |
3 | 48 in. | 24 in. |
Each for $745.84
|
|
|||
Description
- Chrome-plated models are suitable for a variety of dry environments and include 5 in. (127 mm) polyurethane casters with plated caster horns
- Shelf construction makes units easy to assemble and makes relocation of shelves quick and simple
- Open wire construction minimizes dust accumulation, maximizes visibility and air circulation.
Specifications
| Wire Utility Cart | |
| Silver | |
| 39 in. | |
| 900 lb. | |
| Chrome |
| Not Autoclavable | |
| Storage and Transport | |
| 99 cm | |
| 408 kg |