missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Utility Cart with Wired Shelves, Chrome

Carts consist of wire shelves with plastic split sleeves, two one-piece handles, donut bumpers, and casters.

Supplier:  Fisherbrand™ FS1842NC2UC

 View more versions of this product

Catalog No. 13460185

Only null left

Description

Description

  • Chrome-plated models are suitable for a variety of dry environments and include 5 in. (127 mm) polyurethane casters with plated caster horns
  • Shelf construction makes units easy to assemble and makes relocation of shelves quick and simple
  • Open wire construction minimizes dust accumulation, maximizes visibility and air circulation.
Specifications

Chemical Identifiers

Wire Utility Cart
Silver
39 in.
900 lb.
Chrome
Not Autoclavable
Storage and Transport
99 cm
408 kg

Specifications

Not Autoclavable
Silver
Storage and Transport
99 cm
106.7 cm
408 kg
2
33.8 lb.
45.7 cm
Wire Utility Cart
42 x 18 x 39 in.
39 in.
42 in.
900 lb.
Chrome
15 kg
18 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions