missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylthioacetamide, 97%
Description
2-Phenylthioacetamide find extensive use in organic synthesis. N-bromo and N-chloro succinimides are halogenating agents. Phthalimides are used in the synthesis of primary amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Phenylthioacetamide find extensive use in organic synthesis. N-bromo and N-chloro succinimides are halogenating agents. Phthalimides are used in the synthesis of primary amines.
Solubility
Slightly Soluble in water (1.8 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
2-Phenylthioacetamide find extensive use in organic synthesis. N-bromo and N-chloro succinimides are halogenating agents. Phthalimides are used in the synthesis of primary amines.
Solubility
Slightly Soluble in water (1.8 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Spécifications
Spécifications
| Melting Point | 91°C to 95°C |
| Quantity | 250 mg |
| Solubility Information | Slightly Soluble in water (1.8g/L) (25°C). |
| Formula Weight | 151.23 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Phenylthioacetamide |
Sécurité et manipulation
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only