missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthaldehyde, 98%, Thermo Scientific™

Identifiants chimiques

CAS 66-99-9
Molecular Formula C11H8O
Molecular Weight (g/mol) 156.18
MDL Number MFCD00004094
InChI Key PJKVFARRVXDXAD-UHFFFAOYSA-N
Synonym 2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene
PubChem CID 6201
ChEBI CHEBI:52368
IUPAC Name naphthalene-2-carbaldehyde
SMILES C1=CC=C2C=C(C=CC2=C1)C=O
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC128100050
FDS Afficher les documents
Thermo Scientific Chemicals
128100050
5g Glass bottle N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
AC128100250
FDS Afficher les documents
Thermo Scientific Chemicals
128100250
N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
AC128101000
FDS Afficher les documents
Thermo Scientific Chemicals
128101000
100g Glass bottle N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Identifiants chimiques

66-99-9
156.18
PJKVFARRVXDXAD-UHFFFAOYSA-N
6201
naphthalene-2-carbaldehyde
C11H8O
MFCD00004094
2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene
CHEBI:52368
C1=CC=C2C=C(C=CC2=C1)C=O

Spécifications

66-99-9
100.0
>100°C
97.5% min. (GC)
C11H8O
MFCD00004094
07, 401
Solubility in water: slightly soluble. Other solubilities: soluble in hot water
C1=CC=C2C=C(C=CC2=C1)C=O
156.18
CHEBI:52368
98%
97.5
160.0°C (19.0 mmHg)
Authentic
Glass bottle
C10H7CHO
5g
2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene
PJKVFARRVXDXAD-UHFFFAOYSA-N
naphthalene-2-carbaldehyde
6201
156.18
2-Naphthaldehyde
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 200-640-2

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only