Learn More
2-Methylbutane, for HPLC
CAS: 78-78-4 | C5H12 | 72.15 g/mol
$399.26 - $817.01
Identifiants chimiques
| CAS | 78-78-4 |
|---|---|
| Molecular Formula | C5H12 |
| Molecular Weight (g/mol) | 72.15 |
| MDL Number | MFCD00009338 |
| InChI Key | QWTDNUCVQCZILF-UHFFFAOYSA-N |
| Synonym | isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 |
| PubChem CID | 6556 |
| ChEBI | CHEBI:30362 |
| IUPAC Name | 2-methylbutane |
| SMILES | CCC(C)C |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC427730010
|
Thermo Scientific Chemicals
427730010 |
1 L | Glass bottle |
chaque for $399.26
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC427730025
|
Thermo Scientific Chemicals
427730025 |
2.5 L | Glass bottle |
chaque for $817.01
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for HPLCIdentifiants chimiques
| 78-78-4 | |
| 72.15 | |
| QWTDNUCVQCZILF-UHFFFAOYSA-N | |
| 6556 | |
| 2-methylbutane |
| C5H12 | |
| MFCD00009338 | |
| isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
| CHEBI:30362 | |
| CCC(C)C |
Spécifications
| 78-78-4 | |
| -160.0°C | |
| 30.0°C | |
| Glass bottle | |
| 1.3530 to 1.3550 | |
| MFCD00009338 | |
| 0.0003% max. | |
| isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
| QWTDNUCVQCZILF-UHFFFAOYSA-N | |
| 2-methylbutane | |
| 6556 | |
| 72.15 | |
| HPLC | |
| 2-Methylbutane, For HPLC |
| 0.01 max. 1 cm cell vs HPLC−grade water at 240nm, 0.01 max. 1 cm cell vs HPLC−grade water at 400nm, 0.07 max. 1 cm cell vs HPLC−grade water at 220nm, 0.3 max. 1 cm cell vs HPLC−grade water at 210nm, 1 max. 1 cm cell vs HPLC grade water at 195nm | |
| 0.6200g/mL | |
| −51°C | |
| C5H12 | |
| C2H5CH(CH3)2 | |
| 1 L | |
| 0.62 | |
| Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons, oils and ether, slightly soluble in alcohol | |
| CCC(C)C | |
| 72.15 | |
| CHEBI:30362 | |
| ≥99.0% | |
| Liquid |
Sécurité et manipulation
GHS H Statement
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
Repeated exposure may cause skin drynes
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flam
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-142-8
RUO – Research Use Only