missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-pyrroline, Thermo Scientific Chemicals

$488.99 - $488.99

Identifiants chimiques

CAS 872-32-2
Molecular Formula C5H10N
Molecular Weight (g/mol) 84.14
MDL Number MFCD00128806
InChI Key OZLJZWIVBYLSOK-UHFFFAOYSA-N
Synonym 2-methyl-1-pyrroline, 3,4-dihydro-5-methyl-2h-pyrrole, 1pyrroline2methyl, 2h-pyrrole, 3,4-dihydro-5-methyl, pubchem9412
PubChem CID 70103
ChEBI CHEBI:78856
IUPAC Name 5-methyl-3,4-dihydro-2H-pyrrole
SMILES C[C+]1CCCN1
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AA44853AC
Afficher les documents
Thermo Scientific Chemicals
044853AC
25 mL
chaque for $488.99
Il en reste null
 
Description

Description

2-Methyl-1-pyrroline acts as monocyclic imine. It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

872-32-2
84.14
OZLJZWIVBYLSOK-UHFFFAOYSA-N
70103
5-methyl-3,4-dihydro-2H-pyrrole
C5H10N
MFCD00128806
2-methyl-1-pyrroline, 3,4-dihydro-5-methyl-2h-pyrrole, 1pyrroline2methyl, 2h-pyrrole, 3,4-dihydro-5-methyl, pubchem9412
CHEBI:78856
C[C+]1CCCN1

Spécifications

872-32-2
104°C to 105°C
C5H10N
25 mL
2-methyl-1-pyrroline, 3,4-dihydro-5-methyl-2h-pyrrole, 1pyrroline2methyl, 2h-pyrrole, 3,4-dihydro-5-methyl, pubchem9412
OZLJZWIVBYLSOK-UHFFFAOYSA-N
5-methyl-3,4-dihydro-2H-pyrrole
70103
83.13
2-Methyl-1-pyrroline
0.878 g/mL
10°C (50°F)
MFCD00128806
UN2733
Soluble in water.
C[C+]1CCCN1
84.14
CHEBI:78856
Liquid
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 212-822-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only