missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Supplier:  TCI America A140525ML

 Afficher plus de versions de ce produit

Numéro de catalogue. A140525ML


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

2-Methoxyethyl Acrylate (stabilized with MEHQ)
Yellow
C6H10O3
25 mL
2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca
COCCOC(=O)C=C
130.14
130.14
Liquid
3121-61-7
56°C
MFCD00048149,MFCD00803685
1992
HFCUBKYHMMPGBY-UHFFFAOYSA-N
2-methoxyethyl prop-2-enoate
18392
≥98.0% (GC)

Identifiants chimiques

3121-61-7
130.14
HFCUBKYHMMPGBY-UHFFFAOYSA-N
18392
COCCOC(=O)C=C
C6H10O3
MFCD00048149,MFCD00803685
2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca
2-methoxyethyl prop-2-enoate
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : (2)-1004

missing translation for 'rtecsNumber' : KL6000000

missing translation for 'tsca' : Yes

FDS
Documents

Documents

Promotions

Promotions