missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptophenol, 98+%

Numéro de catalogue. p-3586348
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Packaging:
Glass bottle
Numéro de catalogue. Quantity Packaging
AC385600010 1 g Glass bottle
1 options

Numéro de catalogue. AC385600010

Supplier: Thermo Scientific Chemicals 385600010

Il en reste null

CAS: 1121-24-0 | C6H6OS | 126.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 1121-24-0
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.18
MDL Number MFCD00040447
InChI Key VMKYTRPNOVFCGZ-UHFFFAOYSA-N
Synonym 2-hydroxythiophenol, 2-mercaptophenol, o-mercaptophenol, phenol, 2-mercapto, 2-hydroxy thiophenol, 2-hydroxybenzenethiol, o-hydroxythiophenol, o-hydroxybenzenothiol, 2-hydroxybenzenothiol, mercaptophenol
PubChem CID 70718
IUPAC Name 2-sulfanylphenol
SMILES C1=CC=C(C(=C1)O)S

Spécifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 20.0°C
Color Green to Yellow
Density 1.2200g/mL
Boiling Point 40.0°C (300.0 mmHg)
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.6100 to 1.6120
Linear Formula SHC6H4OH
Quantity 1 g
Specific Gravity 1.22
Solubility Information Solubility in water: slightly soluble.
Formula Weight 126.18
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Mercaptophenol
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-326-8

RUO – Research Use Only