missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ 2-Mercaptoethanol (50 mM)

Catalog No. 501147851
Encompass
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501147851 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501147851 Supplier Gibco™ Supplier No. 31350010

2-Mercaptoethanol (50 mM)

Gibco™ 2-Mercaptoethanol (also known as beta-mercaptoethanol or BME) is a potent reducing agent used in cell culture medium to prevent toxic levels of oxygen radicals. Mercaptoethanol is not stable in solution so most protocols require daily supplementation. Gibco™ 2-Mercaptoethanol is made at 50 mM in Dulbecco's Phosphate Buffered Saline (DPBS).

Product Use
For Research Use Only. Not for use in diagnostic procedures.

cGMP Manufacturing and Quality System
Gibco™ 2-Mercaptoethanol is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Specifications

Content And Storage Storage conditions: 2 – 8°C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Cell Type Mammalian Cells
Concentration 50 mM
Form Liquid
Product Type BME
pH 6 to 8
Quantity 20 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.