Learn More
2-Hydroxyethyl methacrylate, 97%, stabilized
CAS: 868-77-9 | C6H10O3 | 130.14 g/mol
Supplier: Thermo Scientific Chemicals 156330025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| 2-Hydroxyethyl methacrylate | |
| 2mg KOH/g max. | |
| 100.0 | |
| 1.0730g/mL | |
| 106°C | |
| 96% min. (GC) | |
| C6H10O3 | |
| H2C=C(CH3)CO2CH2CH2OH | |
| 2.5 kg | |
| 1.073 | |
| Solubility in water: soluble. Other solubilities: soluble in most common organic solvents | |
| CC(=C)C(=O)OCCO | |
| 130.14 | |
| 13360 | |
| CHEBI:34288 | |
| 97% |
| 868-77-9,150-76-5 | |
| 96.0 | |
| -12.0°C | |
| 213°C | |
| Authentic | |
| Glass bottle | |
| 1.4510 to 1.4530 | |
| MFCD00002863 | |
| 02,III,1291 | |
| 2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1 | |
| WOBHKFSMXKNTIM-UHFFFAOYSA-N | |
| 2-hydroxyethyl 2-methylprop-2-enoate | |
| 200 to 400ppm MEHQ | |
| 5 mPa.s (30°C) | |
| 130.14 | |
| Liquid |
Identifiants chimiques
| 868-77-9 | |
| 130.14 | |
| WOBHKFSMXKNTIM-UHFFFAOYSA-N | |
| 13360 | |
| 2-hydroxyethyl 2-methylprop-2-enoate |
| C6H10O3 | |
| MFCD00002863 | |
| 2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1 | |
| CHEBI:34288 | |
| CC(=C)C(=O)OCCO |
Sécurité et manipulation
GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 212-782-2
RUO – Research Use Only