missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoroacetophenone, 97%

CAS: 445-27-2 | C8H7FO | 138.141 g/mol

Supplier:  Thermo Scientific Chemicals A1512814

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA1512814


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

2'-Fluoroacetophenone is used as starting reagent in the synthesis of ascididemin. It is used to produce 1-(2-piperidin-1-yl-phenyl)-ethanone by reaction with piperidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoroacetophenone is used as starting reagent in the synthesis of ascididemin. It is used to produce 1-(2-piperidin-1-yl-phenyl)-ethanone by reaction with piperidine.

Solubility
Soluble in Acetone, Chloroform, Dichloromethane, Ethanol, Ethyl Acetate and Methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Spécifications

Spécifications

2'-Fluoroacetophenone
1.137
61°C (141°F)
1.5075
25 g
2'-fluoroacetophenone, 1-2-fluorophenyl ethanone, o-fluoroacetophenone, 1-2-fluorophenyl ethan-1-one, 1-acetyl-2-fluorobenzene, ethanone, 1-fluorophenyl, 2-fluoroacetophenone, 2'fluoroacetophenone, 2;-fluoroacetophenone
QMATYTFXDIWACW-UHFFFAOYSA-N
1-(2-fluorophenyl)ethanone
96744
97%
445-27-2
82°C to 83°C (10 mmHg)
C8H7FO
MFCD00000320
2041344
Soluble in Acetone,Chloroform,Dichloromethane,Ethanol,Ethyl Acetate and Methanol.
CC(=O)C1=CC=CC=C1F
138.141
138.14

Identifiants chimiques

445-27-2
138.141
QMATYTFXDIWACW-UHFFFAOYSA-N
96744
CC(=O)C1=CC=CC=C1F
C8H7FO
MFCD00000320
2'-fluoroacetophenone, 1-2-fluorophenyl ethanone, o-fluoroacetophenone, 1-2-fluorophenyl ethan-1-one, 1-acetyl-2-fluorobenzene, ethanone, 1-fluorophenyl, 2-fluoroacetophenone, 2'fluoroacetophenone, 2;-fluoroacetophenone
1-(2-fluorophenyl)ethanone
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319-H335

missing translation for 'einecsNumber' : 207-156-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only