missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Decanone 99.0+%, TCI America™
Supplier: TCI America D164325ML
Spécifications
| 2-Decanone | |
| 4°C | |
| 211°C | |
| MFCD00009571 | |
| 2-decanone, methyl octyl ketone, methyl n-octyl ketone, octyl methyl ketone, unii-gx543olt0r, n-c8h17coch3, gx543olt0r, chembl47127 | |
| CCCCCCCCC(=O)C | |
| 156.269 | |
| CHEBI:77929 | |
| ≥99.0% (GC) |
| 693-54-9 | |
| Colorless | |
| C10H20O | |
| 25 mL | |
| ZAJNGDIORYACQU-UHFFFAOYSA-N | |
| decan-2-one | |
| 12741 | |
| 156.27 | |
| Liquid |
Identifiants chimiques
| 693-54-9 | |
| 156.269 | |
| ZAJNGDIORYACQU-UHFFFAOYSA-N | |
| 12741 | |
| decan-2-one |
| C10H20O | |
| MFCD00009571 | |
| 2-decanone, methyl octyl ketone, methyl n-octyl ketone, octyl methyl ketone, unii-gx543olt0r, n-c8h17coch3, gx543olt0r, chembl47127 | |
| CHEBI:77929 | |
| CCCCCCCCC(=O)C |
Sécurité et manipulation
EINECSNumber : (2)-0542
RTECSNumber : HE0725000
TSCA : Yes