missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl isothiocyanate, 97%

CAS: 2740-81-0 | C7H4ClNS | 169.626 g/mol

Supplier:  Thermo Scientific Chemicals A1823814

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA1823814


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

2-Chlorophenyl isothiocyanate
1.287
25 g
1.6608
MFCD00004801
2081473
DASSPOJBUMBXLU-UHFFFAOYSA-N
1-chloro-2-isothiocyanatobenzene
123171
97%
2740-81-0
260°C to 262°C
Pungent
C7H4ClNS
UN2922
2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, 1-chloro-2-isothiocyanato-benzene, isothiocyanic acid 2-chlorophenyl ester, 2-chlorobenzenisothiocyanate, acmc-1ccw5, o-chlorophenylisothiocyanate, o-chlorophenyl isothiocyanate
C1=CC=C(C(=C1)N=C=S)Cl
169.626
169.64

Identifiants chimiques

2740-81-0
169.626
DASSPOJBUMBXLU-UHFFFAOYSA-N
123171
C1=CC=C(C(=C1)N=C=S)Cl
C7H4ClNS
MFCD00004801
2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, 1-chloro-2-isothiocyanato-benzene, isothiocyanic acid 2-chlorophenyl ester, 2-chlorobenzenisothiocyanate, acmc-1ccw5, o-chlorophenylisothiocyanate, o-chlorophenyl isothiocyanate
1-chloro-2-isothiocyanatobenzene
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H334-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only