missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl Vinyl Ether (stabilized with MEHQ + Triethanolamine) 97.0+%, TCI America™

$247.92 - $1888.29

Identifiants chimiques

CAS 110-75-8
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.549
MDL Number MFCD00000973
InChI Key DNJRKFKAFWSXSE-UHFFFAOYSA-N
Synonym 2-chloroethyl vinyl ether, 2-chloroethoxy ethene, ethene, 2-chloroethoxy, 2-vinyloxyethyl chloride, vinyl 2-chloroethyl ether, ether, 2-chloroethyl vinyl, rcra waste number u042, beta-chloroethyl vinyl ether, 2-chlorethyl vinyl ether
PubChem CID 8074
IUPAC Name 1-chloro-2-ethenoxyethane
SMILES C=COCCCl
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
C017425ML
Afficher les documents
TCI America
C017425ML
25 mL
chaque for $247.92
Il en reste null
 
C0174500ML
Afficher les documents
TCI America
C0174500ML
500 mL
chaque for $1,888.29
Il en reste null
 
Description

Description

Spécifications

Identifiants chimiques

110-75-8
106.549
DNJRKFKAFWSXSE-UHFFFAOYSA-N
8074
C=COCCCl
C4H7ClO
MFCD00000973
2-chloroethyl vinyl ether, 2-chloroethoxy ethene, ethene, 2-chloroethoxy, 2-vinyloxyethyl chloride, vinyl 2-chloroethyl ether, ether, 2-chloroethyl vinyl, rcra waste number u042, beta-chloroethyl vinyl ether, 2-chlorethyl vinyl ether
1-chloro-2-ethenoxyethane

Spécifications

110-75-8
Yellow
C4H7ClO
25 mL
2-chloroethyl vinyl ether, 2-chloroethoxy ethene, ethene, 2-chloroethoxy, 2-vinyloxyethyl chloride, vinyl 2-chloroethyl ether, ether, 2-chloroethyl vinyl, rcra waste number u042, beta-chloroethyl vinyl ether, 2-chlorethyl vinyl ether
C=COCCCl
106.549
106.55
Liquid
-70°C
108°C
MFCD00000973
1992
DNJRKFKAFWSXSE-UHFFFAOYSA-N
1-chloro-2-ethenoxyethane
8074
≥97.0% (GC)
2-Chloroethyl Vinyl Ether (stabilized with MEHQ + Triethanolamine)
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : (2)-0377

missing translation for 'rtecsNumber' : KN6300000

missing translation for 'tsca' : Yes

FDS
missing translation for 'documents'

missing translation for 'documents'