missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Bromomethyl)benzonitrile, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Melting Point | 70°C to 74°C |
| Quantity | 25 g |
| UN Number | UN2923 |
| Beilstein | 742605 |
| Formula Weight | 196.05 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-(Bromomethyl)benzonitrile |
Sécurité et manipulation
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H314-H318-H302-H312-H332-H317 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. May cause an allergic skin reaction. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only