missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methylpyridine 98.0+%, TCI America™

Numéro de catalogue. p-7119490
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B21135G 5 g
B211325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B21135G Fournisseur TCI America Code fournisseur B21135G
Il en reste null

Identifiants chimiques

CAS 3510-66-5
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.025
MDL Number MFCD00209553
InChI Key YWNJQQNBJQUKME-UHFFFAOYSA-N
Synonym 2-bromo-5-picoline, 6-bromo-3-picoline, 2-bromo-5-methyl-pyridine, pyridine, 2-bromo-5-methyl, 2-bromo-5-methyl pyridine, 5-methyl-2-bromopyridine, 2-brom-5-methylpyridin, pubchem1185
PubChem CID 564216
IUPAC Name 2-bromo-5-methylpyridine
SMILES CC1=CN=C(C=C1)Br

Spécifications

Melting Point 44°C
Quantity 5 g
Formula Weight 172.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-methylpyridine