missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Bis[(trimethylsilyl)ethynyl]thieno[3,2-b]thiophene 98.0+%, TCI America™
Supplier: TCI America B51961G
Spécifications
| 2,5-Bis[(trimethylsilyl)ethynyl]thieno[3,2-b]thiophene | |
| 213°C | |
| C16H20S2Si2 | |
| 1 g | |
| C[Si](C)(C)C#CC1=CC2=C(S1)C=C(S2)C#C[Si](C)(C)C | |
| 332.63 | |
| 332.63 | |
| Crystalline Powder |
| 139896-65-4 | |
| Yellow | |
| MFCD29089350 | |
| OWXZBWIZKNUJOL-UHFFFAOYSA-N | |
| trimethyl(2-{5-[2-(trimethylsilyl)ethynyl]thieno[3,2-b]thiophen-2-yl}ethynyl)silane | |
| 85633934 | |
| ≥98.0% (GC) |
Identifiants chimiques
| 139896-65-4 | |
| 332.63 | |
| OWXZBWIZKNUJOL-UHFFFAOYSA-N | |
| trimethyl(2-{5-[2-(trimethylsilyl)ethynyl]thieno[3,2-b]thiophen-2-yl}ethynyl)silane |
| C16H20S2Si2 | |
| MFCD29089350 | |
| 85633934 | |
| C[Si](C)(C)C#CC1=CC2=C(S1)C=C(S2)C#C[Si](C)(C)C |
Sécurité et manipulation
missing translation for 'tsca' : No