missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyridine, 99+%

CAS: 366-18-7 | C10H8N2 | 156.19 g/mol

Supplier:  Thermo Scientific Chemicals A1578236

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA1578236


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Reagent for iron determinations.2,2'-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for iron determinations.2,2′-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.

Solubility
Soluble in organic solvents and slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition.
Spécifications

Spécifications

2,2'-Bipyridine
70°C
121°C (249°F)
MFCD00006212
UN2811
14,3347
Soluble in organic solvents and slightly soluble in water.
C1=CC=C(N=C1)C1=CC=CC=N1
156.19
CHEBI:30351
≥99%
366-18-7
272°C to 273°C
C10H8N2
500 g
113089
2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
ROFVEXUMMXZLPA-UHFFFAOYSA-N
2-pyridin-2-ylpyridine
1474
156.19

Identifiants chimiques

366-18-7
156.19
ROFVEXUMMXZLPA-UHFFFAOYSA-N
1474
C1=CC=C(N=C1)C1=CC=CC=N1
C10H8N2
MFCD00006212
2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
CHEBI:30351
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H301-H312
Toxic if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P310-P302+P352-P312-P330-P361-P363-P501c

H301-H312-H500

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 206-674-4

missing translation for 'rtecsNumber' : DW1750000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only