Learn More
Biotium Inc 10X Fish Gelatin Blocking Agent DFS Item
Supplier:  Biotium 22010DUP
10X Fish Gelatin Blocking Agent is a solution of cold water fish skin gelatin that can be added to blocking buffers to minimize non-specific antibody binding in immunodetection procedures such as Western Blotting and immunostaining. Unlike BSA or milk, fish gelatin does not contain IgG or serum proteins that could cross-react with mammalian antibodies. Fish gelatin can be used to block non-specific binding sites on positively charged nylon or PVDF membranes or used for blocking and dilution of antibodies during immunostaining procedures.
Numéro de catalogue. NC0382999
Il en reste null
Découvrez plus d'offres spéciales