missing translation for 'onlineSavingsMsg'
Learn More

10-(2-Naphthyl)anthracene-9-boronic Acid (contains varying amounts of Anhydride) 98.0+%, TCI America™

Supplier:  TCI America N09291G

Numéro de catalogue. N09291G


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

10-(2-Naphthyl)anthracene-9-boronic Acid (contains varying amounts of Anhydride)
Yellow
MFCD11973625
10-2-naphthyl anthracene-9-boronic acid, 10-naphthalen-2-yl anthracen-9-yl boronic acid, boronic acid, 10-2-naphthalenyl-9-anthracenyl, 10-naphthalen-2-yl anthracen-9-ylboronic acid, 10-2-naphthyl-9-anthrylboronic acid, 10-2-naphthyl-9-anthryl boronic acid, 10-naphth-2-ylanthracen-9-ylboronic acid, 10-naphthalen-2-yl-anthracene-9-boronic acid, 10-naphthalene-2-yl-anthracene-9-boronic acid
B(C1=C2C=CC=CC2=C(C3=CC=CC=C13)C4=CC5=CC=CC=C5C=C4)(O)O
348.208
348.21
Crystalline Powder
597554-03-5
C24H17BO2
1 g
YGVDBZMVEURVOW-UHFFFAOYSA-N
(10-naphthalen-2-ylanthracen-9-yl)boronic acid
23088556
≥98.0% (HPLC)

Identifiants chimiques

597554-03-5
348.208
YGVDBZMVEURVOW-UHFFFAOYSA-N
23088556
B(C1=C2C=CC=CC2=C(C3=CC=CC=C13)C4=CC5=CC=CC=C5C=C4)(O)O
C24H17BO2
MFCD11973625
10-2-naphthyl anthracene-9-boronic acid, 10-naphthalen-2-yl anthracen-9-yl boronic acid, boronic acid, 10-2-naphthalenyl-9-anthracenyl, 10-naphthalen-2-yl anthracen-9-ylboronic acid, 10-2-naphthyl-9-anthrylboronic acid, 10-2-naphthyl-9-anthryl boronic acid, 10-naphth-2-ylanthracen-9-ylboronic acid, 10-naphthalen-2-yl-anthracene-9-boronic acid, 10-naphthalene-2-yl-anthracene-9-boronic acid
(10-naphthalen-2-ylanthracen-9-yl)boronic acid
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'tsca' : No

FDS
Documents

Documents

Promotions

Promotions