missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole 98.0+%, TCI America™

$78.29 - $223.68

Chemical Identifiers

CAS 552846-17-0
Molecular Formula C14H23BN2O4
Molecular Weight (g/mol) 294.16
MDL Number MFCD05663873
InChI Key IPISOFJLWYBCAV-UHFFFAOYSA-N
Synonym 1-boc-pyrazole-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-carboxylate, 1-boc-4-pyrazoleboronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole-1-carboxylate, n-boc-pyrazole-4-boronic acid pinacol ester, 1-tert-butoxycarbonyl-1h-pyrazole-4-boronic acid, pinacol ester, n-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrazole, 1-tert-butoxycarbonyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-tert-butoxycarbonyl-4-1h-pyrazoleboronic acid
PubChem CID 16217654
IUPAC Name tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B52031G
View Documents
TCI America
B52031G
1 g
Each for $78.29
Only null left
 
B52035G
View Documents
TCI America
B52035G
5 g
Each for $223.68
Only null left
 
Description

Description

Specifications

Chemical Identifiers

552846-17-0
294.16
IPISOFJLWYBCAV-UHFFFAOYSA-N
16217654
CC(C)(C)OC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
C14H23BN2O4
MFCD05663873
1-boc-pyrazole-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-carboxylate, 1-boc-4-pyrazoleboronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole-1-carboxylate, n-boc-pyrazole-4-boronic acid pinacol ester, 1-tert-butoxycarbonyl-1h-pyrazole-4-boronic acid, pinacol ester, n-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrazole, 1-tert-butoxycarbonyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-tert-butoxycarbonyl-4-1h-pyrazoleboronic acid
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate

Specifications

552846-17-0
White
MFCD05663873
1-boc-pyrazole-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-carboxylate, 1-boc-4-pyrazoleboronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole-1-carboxylate, n-boc-pyrazole-4-boronic acid pinacol ester, 1-tert-butoxycarbonyl-1h-pyrazole-4-boronic acid, pinacol ester, n-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrazole, 1-tert-butoxycarbonyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-tert-butoxycarbonyl-4-1h-pyrazoleboronic acid
CC(C)(C)OC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
294.16
294.16
Crystalline Powder
86°C
C14H23BN2O4
1 g
IPISOFJLWYBCAV-UHFFFAOYSA-N
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate
16217654
≥98.0% (T)
1-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'