missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-cyclopropanecarbonitrile, 97%

CAS: 935-44-4 | C10H9N | 143.19 g/mol

$158.19 - $477.10

Identifiants chimiques

CAS 935-44-4
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00001270
InChI Key ZHFURHRJUWYDKG-UHFFFAOYSA-N
Synonym 1-phenylcyclopropanecarbonitrile, 1-phenyl-1-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-phenyl, 1-cyano-1-phenylcyclopropane, 1-phenyl-cyclopropanecarbonitrile, 1-phenylcyclopropyl cyanide, phenylcyclopropanenitrile, 1-phenylcyclopropanenitrile, phenylcyclopropanecarbonitrile, 1-phenylcyclopropane carbonitrile
PubChem CID 70288
IUPAC Name 1-phenylcyclopropane-1-carbonitrile
SMILES N#CC1(CC1)C1=CC=CC=C1
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC170590100
FDS Afficher les documents
Thermo Scientific Chemicals
170590100
10 g Glass bottle
chaque for $158.19
Il en reste null
 
AC170590500
FDS Afficher les documents
Thermo Scientific Chemicals
170590500
50 g Glass bottle
chaque for $477.10
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

935-44-4
143.19
ZHFURHRJUWYDKG-UHFFFAOYSA-N
70288
N#CC1(CC1)C1=CC=CC=C1
C10H9N
MFCD00001270
1-phenylcyclopropanecarbonitrile, 1-phenyl-1-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-phenyl, 1-cyano-1-phenylcyclopropane, 1-phenyl-cyclopropanecarbonitrile, 1-phenylcyclopropyl cyanide, phenylcyclopropanenitrile, 1-phenylcyclopropanenitrile, phenylcyclopropanecarbonitrile, 1-phenylcyclopropane carbonitrile
1-phenylcyclopropane-1-carbonitrile

Spécifications

935-44-4
100.0
1.0000g/mL
>110°C
96% min. (GC)
C10H9N
C6H5C3H4CN
10 g
1-phenylcyclopropanecarbonitrile, 1-phenyl-1-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-phenyl, 1-cyano-1-phenylcyclopropane, 1-phenyl-cyclopropanecarbonitrile, 1-phenylcyclopropyl cyanide, phenylcyclopropanenitrile, 1-phenylcyclopropanenitrile, phenylcyclopropanecarbonitrile, 1-phenylcyclopropane carbonitrile
N#CC1(CC1)C1=CC=CC=C1
143.19
143.19
Liquid
96.0
Colorless to Yellow
133.0°C to 137.0°C (30.0 mmHg)
Authentic
Glass bottle
1.5370 to 1.5400
MFCD00001270
1
ZHFURHRJUWYDKG-UHFFFAOYSA-N
1-phenylcyclopropane-1-carbonitrile
70288
97%
1-Phenyl-1-cyclopropanecarbonitrile, 97%
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 213-304-5

missing translation for 'tsca' : TSCA

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only