missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyl-3-methylimidazolium hexafluorophosphate, 99%

CAS: 304680-35-1 | C10H19F6N2P | 312.24 g/mol

$190.66 - $614.33

Identifiants chimiques

CAS 304680-35-1
Molecular Formula C10H19F6N2P
Molecular Weight (g/mol) 312.24
MDL Number MFCD03093296
InChI Key YPWSSSRXUOQNMQ-UHFFFAOYSA-N
Synonym 1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate
PubChem CID 2734175
SMILES F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAH2727206
Afficher les documents
Thermo Scientific Chemicals
H2727206
5 g
chaque for $190.66
Il en reste null
 
AAH2727218
Afficher les documents
Thermo Scientific Chemicals
H2727218
50 g
chaque for $614.33
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

304680-35-1
312.24
YPWSSSRXUOQNMQ-UHFFFAOYSA-N
2734175
C10H19F6N2P
MFCD03093296
1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate
F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1

Spécifications

304680-35-1
1.419
5 g
YPWSSSRXUOQNMQ-UHFFFAOYSA-N
1-hexyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
2734175
99%
C10H19F6N2P
MFCD03093296
1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate
F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1
312.24
312.24
1-n-Hexyl-3-methylimidazolium hexafluorophosphate
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only