Learn More
Applied Biosystems™ 1- Methylimidazole/THF, Capping Formula B
Contains 450 mL 1- Methylimidazole/THF, Capping Formula B.
$758.00 - $1318.00
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
4340866
|
Applied Biosystems™
4340866 |
4 x 180 mL |
Each for $758.00
|
|
|||||
401175
|
Applied Biosystems™
401175 |
450 mL |
Each for $1,280.00
|
|
|||||
4340861
|
Applied Biosystems™
4340861 |
4 x 450 mL |
Each for $1,318.00
|
|
|||||
400785
|
Applied Biosystems™
400785 |
180 mL |
Each for $772.00
|
|
|||||
Description
Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.- Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
- Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
- Reagent kits are available for your convenience, and save you time and money.
For Research Use Only. Not for use in diagnostic procedures.