Learn More
Applied Biosystems™ 1- Methylimidazole/THF, Capping Formula B DFS Item
Contains 450 mL 1- Methylimidazole/THF, Capping Formula B.
Supplier:  Applied Biosystems™ 4340861
 View more versions of this product
Catalog No. 4340861
Only null left
Explore available promotions
Description
Description
1-Methylimidazole⁄THF, Capping Formula B, Thermo Scientific™ is a mix of NMI (N-methylimidazole) and THF (tetrahydrofuran).
  • Contains 4 Trap-Pak™ bags filled with 1- Methylimidazole⁄THF, Capping Formula B
  • For research use only; not for use in diagnostics procedures
Specifications
Specifications
4 x 450 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.