missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 1- Methylimidazole/THF, Capping Formula B

Catalog No. 4340866
Encompass
Change view
Click to view available options
Quantity:
180 mL
450 mL
4 x 180 mL
4 x 450 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4340866 4 x 180 mL
401175 450 mL
4340861 4 x 450 mL
400785 180 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 4340866 Supplier Applied Biosystems™ Supplier No. 4340866

Contains 450 mL 1- Methylimidazole/THF, Capping Formula B.

Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.
  • Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
  • Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
  • Reagent kits are available for your convenience, and save you time and money.

Specifications

Quantity 4 x 180 mL

For Research Use Only. Not for use in diagnostic procedures.