Learn More
Applied Biosystems™ 1- Methylimidazole/THF, Capping Formula B DFS Item
Contains 450 mL 1- Methylimidazole/THF, Capping Formula B.
Supplier:  Applied Biosystems™ 401175
 View more versions of this product
Catalog No. 401175
Only null left
Explore available promotions
Description
Description
1-Methylimidazole⁄THF, Capping Formula B, Thermo Scientific™ is a mix of NMI (N-methylimidazole) and THF (tetrahydrofuran).
  • Contains 450mL 1- Methylimidazole⁄THF, Capping Formula B
  • For research use only; not for use in diagnostics procedures
Specifications
Specifications
Shipped at Room Temperature
450 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.