Learn More
Applied Biosystems™ 1- Methylimidazole/THF, Capping Formula B DFS Item
Contains 450 mL 1- Methylimidazole/THF, Capping Formula B.
Supplier:  Applied Biosystems™ 400785
 View more versions of this product
Catalog No. 400785
Only null left
Explore available promotions
Description
Description
Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.
  • Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
  • Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
  • Reagent kits are available for your convenience, and save you time and money.
Specifications
Specifications
Shipped at Room Temperature
180 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.