Learn More
1-Hexene, 98%
Description
1-Hexene is used as a solvent, paint thinner and a medium for conducting chemical reactions. It is a commoner and used in the preparation of both low density and high density polyethylenes. It acts as an adhesives, lubricants, lubricant additives and sealant chemicals. Further, it is used in the preparation of hex-1-en-3-one, heptanal and heptanoic acid. It is also employed in the synthesis of linear plasticizers, oxo-alcohols, motor fuels, automotive additives and biodegradable surfactants. In addition to this, it is used in the preparation of mercaptans, flavors and fragrances, alkyl metals, halides and alkyl silanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Hexene is used as a solvent, paint thinner and a medium for conducting chemical reactions. It is a commoner and used in the preparation of both low density and high density polyethylenes. It acts as an adhesives, lubricants, lubricant additives and sealant chemicals. Further, it is used in the preparation of hex-1-en-3-one, heptanal and heptanoic acid. It is also employed in the synthesis of linear plasticizers, oxo-alcohols, motor fuels, automotive additives and biodegradable surfactants. In addition to this, it is used in the preparation of mercaptans, flavors and fragrances, alkyl metals, halides and alkyl silanes.
Solubility
Miscible with water, ethanol, ethyl ether, benzene and petroleum ether.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
Spécifications
Spécifications
| Melting Point | -140°C |
| Density | 0.673 |
| Boiling Point | 62°C to 64°C |
| Flash Point | −25°C (−13°F) |
| Odor | Mild |
| Refractive Index | 1.388 |
| Linear Formula | CH3(CH2)3CH=CH2 |
| Quantity | 100 mL |
| UN Number | UN2370 |
| Beilstein | 1209240 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H225-H304-H336 |
| Hazard Statement | GHS H Statement H225-H304 Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1-HEXENE |
| EINECSNumber | 209-753-1 |
| RTECSNumber | MP6670000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only