Learn More
1-Allylimidazole, 97%
CAS: 31410-01-2 | C6H8N2 | 108.14 g/mol
$175.64 - $175.64
Identifiants chimiques
| CAS | 31410-01-2 |
|---|---|
| Molecular Formula | C6H8N2 |
| Molecular Weight (g/mol) | 108.14 |
| MDL Number | MFCD00037894 |
| InChI Key | XLXCHZCQTCBUOX-UHFFFAOYSA-N |
| Synonym | 1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole # |
| PubChem CID | 35794 |
| IUPAC Name | 1-prop-2-enylimidazole |
| SMILES | C=CCN1C=CN=C1 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AAL1105318
|
Thermo Scientific Chemicals
L1105318 |
50 g |
chaque for $175.64
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
1-Allylimidazole, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Allylimidazole, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Identifiants chimiques
| 31410-01-2 | |
| 108.14 | |
| XLXCHZCQTCBUOX-UHFFFAOYSA-N | |
| 35794 | |
| C=CCN1C=CN=C1 |
| C6H8N2 | |
| MFCD00037894 | |
| 1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole # | |
| 1-prop-2-enylimidazole |
Spécifications
| 31410-01-2 | |
| 212°C to 213°C | |
| C6H8N2 | |
| MFCD00037894 | |
| UN3267 | |
| 1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole # | |
| XLXCHZCQTCBUOX-UHFFFAOYSA-N | |
| 1-prop-2-enylimidazole | |
| 35794 | |
| 99% |
| 1.003 | |
| 101°C (213°F) | |
| 1.505 | |
| 50 g | |
| 605953 | |
| It is soluble in water. | |
| C=CCN1C=CN=C1 | |
| 108.14 | |
| 108.14 | |
| 1-Allylimidazole |
Sécurité et manipulation
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 250-618-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only