Learn More
1-(4-bromo-1,3-thiazol-2-yl)-4-methylpiperazine, 97%, Thermo Scientific™
Identifiants chimiques
| CAS | 919352-66-2 |
|---|---|
| Molecular Formula | C8H12BrN3S |
| Molecular Weight (g/mol) | 262.17 |
| MDL Number | MFCD09878991 |
| InChI Key | GHLARJPRKBAWMZ-UHFFFAOYSA-N |
| Synonym | 1-4-bromo-1,3-thiazol-2-yl-4-methylpiperazine, 4-bromo-2-4-methylpiperazin-1-yl thiazole, 4-bromo-2-4-methylpiperazin-1-yl-1,3-thiazole, 1-4-bromo-thiazol-2-yl-4-methyl-piperazine, 4-bromo-2-4-methylpiperazinyl-1,3-thiazole, piperazine, 1-4-bromo-2-thiazolyl-4-methyl, 2-4-methylpiperazin-1-yl-4-bromothiazole, 4-bromo-2-n-methylpiperazin-1-yl thiazole |
| PubChem CID | 43811061 |
| IUPAC Name | 4-bromo-2-(4-methylpiperazin-1-yl)-1,3-thiazole |
| SMILES | CN1CCN(CC1)C1=NC(Br)=CS1 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
CC77310DA
|
Thermo Scientific Chemicals
CC77310DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
|
CC77310DE
|
Thermo Scientific Chemicals
CC77310DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Identifiants chimiques
| 919352-66-2 | |
| 262.17 | |
| GHLARJPRKBAWMZ-UHFFFAOYSA-N | |
| 43811061 | |
| CN1CCN(CC1)C1=NC(Br)=CS1 |
| C8H12BrN3S | |
| MFCD09878991 | |
| 1-4-bromo-1,3-thiazol-2-yl-4-methylpiperazine, 4-bromo-2-4-methylpiperazin-1-yl thiazole, 4-bromo-2-4-methylpiperazin-1-yl-1,3-thiazole, 1-4-bromo-thiazol-2-yl-4-methyl-piperazine, 4-bromo-2-4-methylpiperazinyl-1,3-thiazole, piperazine, 1-4-bromo-2-thiazolyl-4-methyl, 2-4-methylpiperazin-1-yl-4-bromothiazole, 4-bromo-2-n-methylpiperazin-1-yl thiazole | |
| 4-bromo-2-(4-methylpiperazin-1-yl)-1,3-thiazole |
Spécifications
| 919352-66-2 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD09878991 | |
| 1-4-bromo-1,3-thiazol-2-yl-4-methylpiperazine, 4-bromo-2-4-methylpiperazin-1-yl thiazole, 4-bromo-2-4-methylpiperazin-1-yl-1,3-thiazole, 1-4-bromo-thiazol-2-yl-4-methyl-piperazine, 4-bromo-2-4-methylpiperazinyl-1,3-thiazole, piperazine, 1-4-bromo-2-thiazolyl-4-methyl, 2-4-methylpiperazin-1-yl-4-bromothiazole, 4-bromo-2-n-methylpiperazin-1-yl thiazole | |
| CN1CCN(CC1)C1=NC(Br)=CS1 | |
| 262.17 | |
| 262.17 | |
| 1-(4-Bromo-1, 3-thiazol-2-yl)-4-methylpiperazine |
| 96.5 | |
| 97% | |
| C8H12BrN3S | |
| 1g | |
| GHLARJPRKBAWMZ-UHFFFAOYSA-N | |
| 4-bromo-2-(4-methylpiperazin-1-yl)-1,3-thiazole | |
| 43811061 | |
| 97% |
Sécurité et manipulation
GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger