missing translation for 'onlineSavingsMsg'
Learn More

1,3-Butanediol, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor B214725LTGL

 Afficher plus de versions de ce produit

Numéro de catalogue. 18608052


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

107-88-0
Glass Bottle
0.99
1
2.5 L
Reagent

Identifiants chimiques

107-88-0
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.